missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,3-dimethylbutan-2-one, Tech., Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD08380EB

 View more versions of this product

Catalog No. CD08380EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-Bromo-3, 3-dimethylbutan-2-one
80.0
1.3300g/mL
(CH3)3CCOCH2Br
25g
SAIRZMWXVJEBMO-UHFFFAOYSA-N
1-bromo-3,3-dimethylbutan-2-one
21642
Tech.
5469-26-1
100.0
C6H11BrO
MFCD00000206
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
CC(C)(C)C(=O)CBr
179.057
179.06
Technical

Chemical Identifiers

5469-26-1
179.057
SAIRZMWXVJEBMO-UHFFFAOYSA-N
21642
CC(C)(C)C(=O)CBr
C6H11BrO
MFCD00000206
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
1-bromo-3,3-dimethylbutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions