Learn More
1-Bromo-3,4,5-trichlorobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6450614
Description
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a multipurpose intermediate commonly used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
1-Bromo-3,4,5-trichlorobenzene | |
White | |
Powder | |
MFCD00155009 | |
3,4,5-trichlorobromobenzene, benzene, 5-bromo-1,2,3-trichloro, 1-bromo-3,4,5-trichlorobenzene, 1-bromo-3,4,5-trichloro-benzene, pubchem23062, acmc-209fpm, ksc497i6l, 1,2,3-trichloro-5-bromobenzene | |
VZUMVBQMJFFYRM-UHFFFAOYSA-N | |
5-bromo-1,2,3-trichlorobenzene | |
4124400 | |
98% |
21928-51-8 | |
25 g | |
C6H2BrCl3 | |
2089937 | |
Slightly soluble in water. | |
ClC1=CC(Br)=CC(Cl)=C1Cl | |
260.34 | |
260.34 |
Chemical Identifiers
21928-51-8 | |
260.34 | |
VZUMVBQMJFFYRM-UHFFFAOYSA-N | |
4124400 | |
ClC1=CC(Br)=CC(Cl)=C1Cl |
C6H2BrCl3 | |
MFCD00155009 | |
3,4,5-trichlorobromobenzene, benzene, 5-bromo-1,2,3-trichloro, 1-bromo-3,4,5-trichlorobenzene, 1-bromo-3,4,5-trichloro-benzene, pubchem23062, acmc-209fpm, ksc497i6l, 1,2,3-trichloro-5-bromobenzene | |
5-bromo-1,2,3-trichlorobenzene |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only