missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,4-dichlorobenzene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 269810250

 View more versions of this product

Catalog No. AC269810250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-3, 4-dichlorobenzene
18282-59-2
100.0
Yellow
Authentic
Glass bottle
1.5986 to 1.6006
MFCD00040849
1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
CFPZDVAZISWERM-UHFFFAOYSA-N
4-bromo-1,2-dichlorobenzene
29013
98+%
98+%
98.0
23.0°C to 24.0°C
140.0°C to 142.0°C (30.0 mmHg)
98+%
C6H3BrCl2
BrC6H3Cl2
25 g
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
ClC1=CC=C(Br)C=C1Cl
225.89
225.91
Liquid After Melting

Chemical Identifiers

18282-59-2
225.89
CFPZDVAZISWERM-UHFFFAOYSA-N
29013
ClC1=CC=C(Br)C=C1Cl
C6H3BrCl2
MFCD00040849
1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
4-bromo-1,2-dichlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-160-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only