missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-fluorobenzene, 95%

CAS: 144584-65-6 | C6H3BrClF | 209.442 g/mol

$97.62 - $330.11

Chemical Identifiers

CAS 144584-65-6
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD03412182
InChI Key DRNWNTAANHEQMK-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
PubChem CID 3685762
IUPAC Name 1-bromo-3-chloro-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6162214
View Documents
Thermo Scientific Chemicals
H6162214
25 g
Each for $97.62
Only null left
 
AAH6162222
View Documents
Thermo Scientific Chemicals
H6162222
100 g
Each for $330.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144584-65-6
209.442
DRNWNTAANHEQMK-UHFFFAOYSA-N
3685762
C1=CC(=C(C(=C1)Br)F)Cl
C6H3BrClF
MFCD03412182
3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
1-bromo-3-chloro-2-fluorobenzene

Specifications

144584-65-6
White to Yellow
MFCD03412182
3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
C1=CC(=C(C(=C1)Br)F)Cl
209.442
209.44
Solid
42°C to 46°C
C6H3BrClF
25 g
DRNWNTAANHEQMK-UHFFFAOYSA-N
1-bromo-3-chloro-2-fluorobenzene
3685762
95%
1-Bromo-3-chloro-2-fluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only