missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-methoxypropane 98.0+%, TCI America™

$39.51 - $88.87

Chemical Identifiers

CAS 36865-41-5
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.02
MDL Number MFCD02258473
InChI Key CEVMYGZHEJSOHZ-UHFFFAOYSA-N
Synonym 3-Bromopropyl Methyl Ether
PubChem CID 524551
IUPAC Name 1-bromo-3-methoxypropane
SMILES COCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34995G
View Documents
TCI America
B34995G
5 g
Each for $39.51
 
B349925G
View Documents
TCI America
B349925G
25 g
Each for $88.87
 
Description

Description

Specifications

Chemical Identifiers

36865-41-5
153.02
CEVMYGZHEJSOHZ-UHFFFAOYSA-N
524551
COCCCBr
C4H9BrO
MFCD02258473
3-Bromopropyl Methyl Ether
1-bromo-3-methoxypropane

Specifications

36865-41-5
132°C
MFCD02258473
1993
CEVMYGZHEJSOHZ-UHFFFAOYSA-N
1-bromo-3-methoxypropane
524551
≥98.0% (GC)
1-Bromo-3-methoxypropane
Yellow
C4H9BrO
5 g
3-Bromopropyl Methyl Ether
COCCCBr
153.02
153.02
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'