missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-methylbutane, 98%, Thermo Scientific Chemicals

$59.73 - $1239.97

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000253
InChI Key YXZFFTJAHVMMLF-UHFFFAOYSA-N
Synonym isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
PubChem CID 7891
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(C)CCBr
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1449222
View Documents
Thermo Scientific Chemicals
A1449222
100 g
Each for $59.73
 
AAA1449230
View Documents
Thermo Scientific Chemicals
A1449230
250 g
Each for $103.18
 
AAA144920B
View Documents
Thermo Scientific Chemicals
A144920B
1000 g
Each for $306.79
 
AAA144920I
View Documents
Thermo Scientific Chemicals
A144920I
5000 g
Each for $1,239.97
 
Description

Description

1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

107-82-4
151.05
YXZFFTJAHVMMLF-UHFFFAOYSA-N
7891
CC(C)CCBr
C5H11Br
MFCD00000253
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
1-bromo-3-methylbutane

Specifications

107-82-4
1.208
22°C (71°F)
1.4409
100 g
773652
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
YXZFFTJAHVMMLF-UHFFFAOYSA-N
1-bromo-3-methylbutane
7891
98%
-112°C
120°C to 121°C
C5H11Br
MFCD00000253
UN2341
14,5114
Not miscible or difficult to mix in water.
CC(C)CCBr
151.05
151.05
1-Bromo-3-methylbutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-BROMO-3-METHYLBUTANE

EINECSNumber : 203-522-9

RTECSNumber : EJ6230000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only