missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-3-nitrobenzene 98.0+%, TCI America™
$47.40 - $446.35
Chemical Identifiers
CAS | 585-79-5 |
---|---|
Molecular Formula | C6H4BrNO2 |
Molecular Weight (g/mol) | 202.01 |
MDL Number | MFCD00024298 |
InChI Key | FWIROFMBWVMWLB-UHFFFAOYSA-N |
Synonym | 3-bromonitrobenzene, m-bromonitrobenzene, benzene, 1-bromo-3-nitro, 3-nitrobromobenzene, m-nitrobromobenzene, 1-bromo-3-nitro-benzene, 3-bromo-1-nitrobenzene, ccris 3114, meta-bromonitrobenzene, bromonitrobenzene-3 |
PubChem CID | 11457 |
IUPAC Name | 1-bromo-3-nitrobenzene |
SMILES | [O-][N+](=O)C1=CC=CC(Br)=C1 |
Chemical Identifiers
585-79-5 | |
202.01 | |
FWIROFMBWVMWLB-UHFFFAOYSA-N | |
11457 | |
[O-][N+](=O)C1=CC=CC(Br)=C1 |
C6H4BrNO2 | |
MFCD00024298 | |
3-bromonitrobenzene, m-bromonitrobenzene, benzene, 1-bromo-3-nitro, 3-nitrobromobenzene, m-nitrobromobenzene, 1-bromo-3-nitro-benzene, 3-bromo-1-nitrobenzene, ccris 3114, meta-bromonitrobenzene, bromonitrobenzene-3 | |
1-bromo-3-nitrobenzene |
Specifications
585-79-5 | |
Yellow | |
C6H4BrNO2 | |
25 g | |
3-bromonitrobenzene, m-bromonitrobenzene, benzene, 1-bromo-3-nitro, 3-nitrobromobenzene, m-nitrobromobenzene, 1-bromo-3-nitro-benzene, 3-bromo-1-nitrobenzene, ccris 3114, meta-bromonitrobenzene, bromonitrobenzene-3 | |
[O-][N+](=O)C1=CC=CC(Br)=C1 | |
202.01 | |
202.01 | |
Crystalline Powder |
55°C | |
257°C | |
MFCD00024298 | |
3459 | |
FWIROFMBWVMWLB-UHFFFAOYSA-N | |
1-bromo-3-nitrobenzene | |
11457 | |
≥98.0% (GC) | |
1-Bromo-3-nitrobenzene |
Safety and Handling
RTECSNumber : CY9040500
TSCA : Yes