missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals

$199.48 - $199.48

Chemical Identifiers

CAS 65423-56-5
Molecular Formula C12H19BrOSi
Molecular Weight (g/mol) 287.272
MDL Number MFCD01318106
InChI Key BFRHMVJJJGUHDI-UHFFFAOYSA-N
Synonym 3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene
PubChem CID 11254705
IUPAC Name (3-bromophenoxy)-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1744006
View Documents
Thermo Scientific Chemicals
L1744006
5 g
Each for $199.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65423-56-5
287.272
BFRHMVJJJGUHDI-UHFFFAOYSA-N
11254705
CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br
C12H19BrOSi
MFCD01318106
3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene
(3-bromophenoxy)-tert-butyl-dimethylsilane

Specifications

66°C (0.3mmHg)
5 g
MFCD01318106
3-bromophenoxy tert-butyl dimethylsilane, 3-bromophenyl tert-butyldimethylsilyl ether, 1-bromo-3-tert-butyldimethylsiloxy benzene, silane, 3-bromophenoxy 1,1-dimethylethyl dimethyl, acmc-1b3kt, 3-t-butyldimethylsilyloxybromobenzene, 3-t-butyldimethylsilyloxy bromobenzene, 3-bromophenoxy-tert-butyldimethylsilane, 3-tert-butyldimethylsiloxy bromobenzene
CC(C)(C)[Si](C)(C)OC1=CC(=CC=C1)Br
287.272
287.27
1.509
1-Bromo-3-(tert-butyldimethylsiloxy)benzene
65423-56-5
C12H19BrOSi
2256298
BFRHMVJJJGUHDI-UHFFFAOYSA-N
(3-bromophenoxy)-tert-butyl-dimethylsilane
11254705
≥98%
Moisture sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only