Learn More
1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
$52.78 - $186.27
Chemical Identifiers
CAS | 106-39-8 |
---|---|
Molecular Formula | C6H4BrCl |
Molecular Weight (g/mol) | 191.45 |
MDL Number | MFCD00000600 |
InChI Key | NHDODQWIKUYWMW-UHFFFAOYSA-N |
Synonym | 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene |
PubChem CID | 7806 |
IUPAC Name | 1-bromo-4-chlorobenzene |
SMILES | ClC1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1216022
|
Thermo Scientific Chemicals
A1216022 |
100 g |
Each for $52.78
|
|
||||||
AAA121600B
|
Thermo Scientific Chemicals
A121600B |
1000 g |
Each for $186.27
|
|
||||||
Description
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
Solubility
Soluble in water 0.045g/L.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
106-39-8 | |
191.45 | |
NHDODQWIKUYWMW-UHFFFAOYSA-N | |
7806 | |
ClC1=CC=C(Br)C=C1 |
C6H4BrCl | |
MFCD00000600 | |
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene | |
1-bromo-4-chlorobenzene |
Specifications
63°C to 67°C | |
1.651 | |
100 g | |
C6H4BrCl | |
UN2811 | |
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene | |
NHDODQWIKUYWMW-UHFFFAOYSA-N | |
1-bromo-4-chlorobenzene | |
7806 | |
≥98% |
106-39-8 | |
196°C | |
Aromatic | |
MFCD00000600 | |
1099014 | |
Soluble in water 0.045g/L. | |
ClC1=CC=C(Br)C=C1 | |
191.45 | |
191.46 | |
1-Bromo-4-chlorobenzene |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 203-392-3
RTECSNumber : CY9006000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only