missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-n-butylbenzene, 98%, Thermo Scientific Chemicals

$139.16 - $374.37

Chemical Identifiers

CAS 41492-05-1
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00040934
InChI Key BRGVKVZXDWGJBX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
PubChem CID 521059
IUPAC Name 1-bromo-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2397914
View Documents
Thermo Scientific Chemicals
B2397914
25 g
Each for $139.16
 
AAB2397922
View Documents
Thermo Scientific Chemicals
B2397922
100 g
Each for $374.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41492-05-1
213.118
BRGVKVZXDWGJBX-UHFFFAOYSA-N
521059
CCCCC1=CC=C(C=C1)Br
C10H13Br
MFCD00040934
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
1-bromo-4-butylbenzene

Specifications

41492-05-1
242°C
C10H13Br
MFCD00040934
2080263
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-butylbenzene
521059
98%
1.208
108°C (226°F)
1.53
25 g
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
CCCCC1=CC=C(C=C1)Br
213.118
213.12
1-Bromo-4-n-butylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only