missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-4-(n-pentyloxy)benzene, Thermo Scientific™
$121.90 - $121.90
Chemical Identifiers
CAS | 30752-18-2 |
---|---|
Molecular Formula | C11H15BrO |
Molecular Weight (g/mol) | 243.14 |
MDL Number | MFCD00156123 |
InChI Key | ILLQRHZDICIFRQ-UHFFFAOYSA-N |
Synonym | 4-n-pentyloxy bromobenzene, 1-bromo-4-pentyloxy benzene, 1-bromo-4-pentoxy-benzene, 1-bromo-4-n-pentyloxy benzene, 4-n-amyloxybromobenzene, 4-n-pentyloxybromobenzene, 1-bromo-4-pentyloxybenzene, 4-bromophenylpentyl ether, 4-n-amyl-oxybromobenzene, para-n-pentyloxybromobenzene |
PubChem CID | 7016905 |
IUPAC Name | 1-bromo-4-pentoxybenzene |
SMILES | CCCCCOC1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1261406
|
Thermo Scientific Chemicals
A1261406 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
30752-18-2 | |
243.14 | |
ILLQRHZDICIFRQ-UHFFFAOYSA-N | |
7016905 | |
CCCCCOC1=CC=C(Br)C=C1 |
C11H15BrO | |
MFCD00156123 | |
4-n-pentyloxy bromobenzene, 1-bromo-4-pentyloxy benzene, 1-bromo-4-pentoxy-benzene, 1-bromo-4-n-pentyloxy benzene, 4-n-amyloxybromobenzene, 4-n-pentyloxybromobenzene, 1-bromo-4-pentyloxybenzene, 4-bromophenylpentyl ether, 4-n-amyl-oxybromobenzene, para-n-pentyloxybromobenzene | |
1-bromo-4-pentoxybenzene |
Specifications
30752-18-2 | |
MFCD00156123 | |
4-n-pentyloxy bromobenzene, 1-bromo-4-pentyloxy benzene, 1-bromo-4-pentoxy-benzene, 1-bromo-4-n-pentyloxy benzene, 4-n-amyloxybromobenzene, 4-n-pentyloxybromobenzene, 1-bromo-4-pentyloxybenzene, 4-bromophenylpentyl ether, 4-n-amyl-oxybromobenzene, para-n-pentyloxybromobenzene | |
CCCCCOC1=CC=C(Br)C=C1 | |
243.14 | |
243.15 |
C11H15BrO | |
5 g | |
ILLQRHZDICIFRQ-UHFFFAOYSA-N | |
1-bromo-4-pentoxybenzene | |
7016905 | |
1-Bromo-4-(n-pentyloxy)benzene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures