Learn More
1-Bromo-4-(trifluoromethylthio)benzene, 97%, Thermo Scientific Chemicals
$67.29 - $205.82
Chemical Identifiers
CAS | 333-47-1 |
---|---|
Molecular Formula | C7H4BrF3S |
Molecular Weight (g/mol) | 257.068 |
MDL Number | MFCD00040840 |
InChI Key | CQQBVMXLTBXNLB-UHFFFAOYSA-N |
Synonym | 4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene |
PubChem CID | 736413 |
IUPAC Name | 1-bromo-4-(trifluoromethylsulfanyl)benzene |
SMILES | C1=CC(=CC=C1SC(F)(F)F)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2087403
|
Thermo Scientific Chemicals
B2087403 |
1 g |
Each for $67.29
|
|
|||||
AAB2087406
|
Thermo Scientific Chemicals
B2087406 |
5 g |
Each for $205.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
333-47-1 | |
257.068 | |
CQQBVMXLTBXNLB-UHFFFAOYSA-N | |
736413 | |
C1=CC(=CC=C1SC(F)(F)F)Br |
C7H4BrF3S | |
MFCD00040840 | |
4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene | |
1-bromo-4-(trifluoromethylsulfanyl)benzene |
Specifications
333-47-1 | |
199°C to 200°C | |
C7H4BrF3S | |
MFCD00040840 | |
2361926 | |
CQQBVMXLTBXNLB-UHFFFAOYSA-N | |
1-bromo-4-(trifluoromethylsulfanyl)benzene | |
736413 | |
97% |
1.71 | |
83°C (181°F) | |
1.517 | |
1 g | |
4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene | |
C1=CC(=CC=C1SC(F)(F)F)Br | |
257.068 | |
257.07 | |
1-Bromo-4-(trifluoromethylthio)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only