missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2047918

 View more versions of this product

Catalog No. AAB2047918


Explore available promotions

Description

Description

1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
Specifications

Specifications

1-Bromo-5-chloropentane
1.408
95°C (203°F)
1.4836
50 g
pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PHHNNDKXQVKJEP-UHFFFAOYSA-N
1-bromo-5-chloropentane
96070
98%
54512-75-3
210°C to 212°C
C5H10BrCl
MFCD00001016
1732341
Insoluble in water.
ClCCCCCBr
185.49
185.5

Chemical Identifiers

54512-75-3
185.49
PHHNNDKXQVKJEP-UHFFFAOYSA-N
96070
ClCCCCCBr
C5H10BrCl
MFCD00001016
pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
1-bromo-5-chloropentane
Safety and Handling

Safety and Handling

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 259-193-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only