missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-6-chlorohexane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2161906

 View more versions of this product

Catalog No. AAB2161906


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-6-chlorohexane
1.337
101°C (214°F)
1.4806
5 g
JTYUIAOHIYZBPB-UHFFFAOYSA-N
1-bromo-6-chlorohexane
80516
97%
6294-17-3
109°C to 110°C (2 mmHg)
C6H12BrCl
MFCD00001019
6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
ClCCCCCCBr
199.52
199.53

Chemical Identifiers

6294-17-3
199.52
JTYUIAOHIYZBPB-UHFFFAOYSA-N
80516
ClCCCCCCBr
C6H12BrCl
MFCD00001019
6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
1-bromo-6-chlorohexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-555-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only