missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoadamantane, 99%, Thermo Scientific™
Identifiants chimiques
| CAS | 768-90-1 |
|---|---|
| Molecular Formula | C10H15Br |
| Molecular Weight (g/mol) | 215.13 |
| MDL Number | MFCD00074721 |
| InChI Key | VQHPRVYDKRESCL-UHFFFAOYSA-N |
| Synonym | 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane |
| PubChem CID | 79106 |
| IUPAC Name | 1-bromoadamantane |
| SMILES | C1C2CC3CC1CC(C2)(C3)Br |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC149030250
|
Thermo Scientific Chemicals
149030250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
AC149031000
|
Thermo Scientific Chemicals
149031000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 768-90-1 | |
| 215.13 | |
| VQHPRVYDKRESCL-UHFFFAOYSA-N | |
| 79106 | |
| C1C2CC3CC1CC(C2)(C3)Br |
| C10H15Br | |
| MFCD00074721 | |
| 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
| 1-bromoadamantane |
Spécifications
| 768-90-1 | |
| 100.0 | |
| 98.5% min. (GC) | |
| C10H15Br | |
| 25g | |
| Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,chloroform,benzene,,methanol and ethanol | |
| C1C2CC3CC1CC(C2)(C3)Br | |
| 215.13 | |
| 215.13 | |
| 1-Bromoadamantane, 99% |
| 98.5 | |
| Authentic | |
| Glass bottle | |
| MFCD00074721 | |
| 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
| VQHPRVYDKRESCL-UHFFFAOYSA-N | |
| 1-bromoadamantane | |
| 79106 | |
| 99% |
Sécurité et manipulation
MOISTURE SENSITIVE
missing translation for 'einecsNumber' : 212-200-7
missing translation for 'tsca' : TSCA
RUO â Research Use Only