Learn More
1-Bromobutane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106770025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Bromobutane | |
98.5 | |
-112.0°C | |
1.2700g/mL | |
10°C | |
98.5% min. (GC) | |
C4H9Br | |
CH3(CH2)3Br | |
2.5 L | |
1.27 | |
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
1-bromobutane | |
8002 | |
99% |
109-65-9 | |
100.0 | |
Colorless to Yellow | |
100.0°C to 104.0°C | |
Authentic | |
Glass bottle | |
1.4380 to 1.4404 | |
MFCD00000260 | |
01,119 | |
15,1555 | |
Solubility in water: 0.608g/L (30°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene and ccl4 | |
CCCCBr | |
137.02 | |
137.02 | |
Liquid |
Chemical Identifiers
109-65-9 | |
137.02 | |
MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
8002 | |
CCCCBr |
C4H9Br | |
MFCD00000260 | |
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
1-bromobutane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 203-691-9
RUO – Research Use Only