missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromobutane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106772500

 View more versions of this product

Catalog No. AC106772500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromobutane
98.5
-112.0°C
1.2700g/mL
10°C
98.5% min. (GC)
C4H9Br
CH3(CH2)3Br
250 mL
1.27
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
MPPPKRYCTPRNTB-UHFFFAOYSA-N
1-bromobutane
8002
99%
109-65-9
100.0
Colorless to Yellow
100.0°C to 104.0°C
Authentic
Glass bottle
1.4380 to 1.4404
MFCD00000260
01,119
15,1555
Solubility in water: 0.608g/L (30°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene and ccl4
CCCCBr
137.02
137.02
Liquid

Chemical Identifiers

109-65-9
137.02
MPPPKRYCTPRNTB-UHFFFAOYSA-N
8002
CCCCBr
C4H9Br
MFCD00000260
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
1-bromobutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 203-691-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only