missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromocarbazole 98.0+%, TCI America™
$59.65 - $171.49
Chemical Identifiers
| CAS | 16807-11-7 |
|---|---|
| Molecular Formula | C12H8BrN |
| Molecular Weight (g/mol) | 246.107 |
| MDL Number | MFCD18450164 |
| InChI Key | VCDOOGZTWDOHEB-UHFFFAOYSA-N |
| PubChem CID | 11230335 |
| IUPAC Name | 1-bromo-9H-carbazole |
| SMILES | C1=CC=C2C(=C1)C3=C(N2)C(=CC=C3)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4816200MG
|
TCI America
B4816200MG |
200 mg |
Each for $59.65
|
|
|||||
|
B48161G
|
TCI America
B48161G |
1 g |
Each for $171.49
|
|
|||||
Chemical Identifiers
| 16807-11-7 | |
| 246.107 | |
| VCDOOGZTWDOHEB-UHFFFAOYSA-N | |
| 1-bromo-9H-carbazole |
| C12H8BrN | |
| MFCD18450164 | |
| 11230335 | |
| C1=CC=C2C(=C1)C3=C(N2)C(=CC=C3)Br |
Specifications
| 16807-11-7 | |
| White-Yellow | |
| MFCD18450164 | |
| VCDOOGZTWDOHEB-UHFFFAOYSA-N | |
| 1-bromo-9H-carbazole | |
| 11230335 | |
| ≥98.0% (GC) | |
| 1-Bromocarbazole |
| 124°C | |
| C12H8BrN | |
| 200 mg | |
| C1=CC=C2C(=C1)C3=C(N2)C(=CC=C3)Br | |
| 246.107 | |
| 246.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No