missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromodecane, 98%, Thermo Scientific Chemicals

$52.22 - $1296.59

Chemical Identifiers

CAS 112-29-8
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.18
MDL Number MFCD00000221
InChI Key MYMSJFSOOQERIO-UHFFFAOYSA-N
Synonym decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
PubChem CID 8173
IUPAC Name 1-bromodecane
SMILES CCCCCCCCCCBr
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1134022
View Documents
Thermo Scientific Chemicals
A1134022
100 g
Each for $52.22
 
AAA1134036
View Documents
Thermo Scientific Chemicals
A1134036
500 g
Each for $108.66
 
AAA113400E
View Documents
Thermo Scientific Chemicals
A113400E
2500 g
Each for $376.19
 
AAA113400C
View Documents
Thermo Scientific Chemicals
A113400C
10,000 g
Each for $1,296.59
 
Description

Description

1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

112-29-8
221.18
MYMSJFSOOQERIO-UHFFFAOYSA-N
8173
CCCCCCCCCCBr
C10H21Br
MFCD00000221
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
1-bromodecane

Specifications

112-29-8
1.067
94°C (201°F)
1.456
MFCD00000221
UN2810
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
MYMSJFSOOQERIO-UHFFFAOYSA-N
1-bromodecane
8173
98%
-30°C
238°C
C10H21Br
CH3(CH2)9Br
100 g
1735227
Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water.
CCCCCCCCCCBr
221.18
221.19
1-Bromodecane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331
Toxic if inhaled.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 203-955-3

RTECSNumber : HD6850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only