missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99+%

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

$143.79 - $2045.39

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC107045000
SDS View Documents
Thermo Scientific Chemicals
107045000
500 mL Glass bottle
Each for $143.79
Only null left
 
AC107040025
SDS View Documents
Thermo Scientific Chemicals
107040025
2.5 L Glass bottle
Each for $568.65
Only null left
 
AC107040100
SDS View Documents
Thermo Scientific Chemicals
107040100
10 L Plastic drum
Each for $2,045.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-25-1
165.07
MNDIARAMWBIKFW-UHFFFAOYSA-N
8101
CCCCCCBr
C6H13Br
MFCD00000271
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
1-bromohexane

Specifications

111-25-1
99.0
-85°C
154°C to 158°C
Authentic
Glass bottle
1.4465 to 1.4485
MFCD00000271
01,144
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
MNDIARAMWBIKFW-UHFFFAOYSA-N
1-bromohexane
8101
99+%
1-Bromohexane
0.1% max. (as HBr)
100.0
1.1700g/mL
57°C
0.5% max. Hexanol (GC)
C6H13Br
CH3(CH2)5Br
500 mL
1.17
Solubility in water: insoluble
CCCCCCBr
165.07
165.07
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only