Learn More
1-Bromonaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1412209
Description
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.
Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
1-Bromonaphthalene | |
-2°C to 0°C | |
279°C to 281°C | |
97% | |
1.657 | |
10 g | |
14,1425 | |
Miscible with chloroform,ethanol,ether,benzene and ethyl acetate. Slightly miscible with water. | |
C1=CC=C2C(=C1)C=CC=C2Br | |
207.07 | |
207.08 | |
Liquid |
90-11-9 | |
1.485 | |
>110°C (230°F) | |
C10H7Br | |
MFCD00003868 | |
1906414 | |
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene | |
DLKQHBOKULLWDQ-UHFFFAOYSA-N | |
1-bromonaphthalene | |
7001 | |
97% |
Chemical Identifiers
90-11-9 | |
207.07 | |
DLKQHBOKULLWDQ-UHFFFAOYSA-N | |
7001 | |
C1=CC=C2C(=C1)C=CC=C2Br |
C10H7Br | |
MFCD00003868 | |
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene | |
1-bromonaphthalene |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 201-965-2
RTECSNumber : QJ1545000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only