Learn More
1-Bromopentane, 99%, Thermo Scientific Chemicals
$78.32 - $823.19
Chemical Identifiers
CAS | 110-53-2 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.047 |
MDL Number | MFCD00000267 |
InChI Key | YZWKKMVJZFACSU-UHFFFAOYSA-N |
Synonym | n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0 |
PubChem CID | 8057 |
IUPAC Name | 1-bromopentane |
SMILES | CCCCCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1082130
|
Thermo Scientific Chemicals
A1082130 |
250 g |
Each for $78.32
|
|
|||||
AAA108210B
|
Thermo Scientific Chemicals
A108210B |
1000 g |
Each for $196.01
|
|
|||||
AAA108210I
|
Thermo Scientific Chemicals
A108210I |
5000 g |
Each for $823.19
|
|
|||||
Description
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.
Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.
Notes
Store away from oxidizing agents and bases.
Chemical Identifiers
110-53-2 | |
151.047 | |
YZWKKMVJZFACSU-UHFFFAOYSA-N | |
8057 | |
CCCCCBr |
C5H11Br | |
MFCD00000267 | |
n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0 | |
1-bromopentane |
Specifications
110-53-2 | |
1.215 | |
31°C (87°F) | |
C5H11Br | |
MFCD00000267 | |
UN1993 | |
14,602 | |
Insoluble in water and soluble in alcohol. Miscible in diethyl ether. | |
CCCCCBr | |
151.047 | |
151.05 | |
1-Bromopentane |
-95°C | |
127°C to 129°C | |
Ether-like | |
1.4436 | |
250 g | |
1730981 | |
n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0 | |
YZWKKMVJZFACSU-UHFFFAOYSA-N | |
1-bromopentane | |
8057 | |
99% |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 203-776-0
RTECSNumber : RZ9770000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only