missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Butanesulfonyl Chloride 98.0+%, TCI America™
$109.38 - $1048.32
Chemical Identifiers
CAS | 2386-60-9 |
---|---|
Molecular Formula | C4H9ClO2S |
Molecular Weight (g/mol) | 156.624 |
MDL Number | MFCD00007464 |
InChI Key | WEDIIKBPDQQQJU-UHFFFAOYSA-N |
Synonym | 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride |
PubChem CID | 75430 |
IUPAC Name | butane-1-sulfonyl chloride |
SMILES | CCCCS(=O)(=O)Cl |
Chemical Identifiers
2386-60-9 | |
156.624 | |
WEDIIKBPDQQQJU-UHFFFAOYSA-N | |
75430 | |
CCCCS(=O)(=O)Cl |
C4H9ClO2S | |
MFCD00007464 | |
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride | |
butane-1-sulfonyl chloride |
Specifications
2386-60-9 | |
65°C | |
MFCD00007464 | |
3265 | |
WEDIIKBPDQQQJU-UHFFFAOYSA-N | |
butane-1-sulfonyl chloride | |
75430 | |
≥98.0% (T) | |
1-Butanesulfonyl Chloride |
Yellow | |
C4H9ClO2S | |
25 g | |
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride | |
CCCCS(=O)(=O)Cl | |
156.624 | |
156.62 | |
Liquid |
Safety and Handling
RTECSNumber : EK5950000
TSCA : Yes