Learn More
1-Butanol (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical BPA3994
Specifications
1-Butanol | |
71-36-3 | |
0.810g/cm³ | |
4 L | |
Glass Bottle | |
Suitable for fluorometric analysis | |
≥99.4 % | |
0.0008mEq/g max. | |
CH3CH2CH2CH2OH | |
6.7mbar at 20°C | |
1120; Colorless | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
≥99.4% |
-89°C | |
Colorless | |
117.6°C | |
Pass Test | |
Liquid | |
0.005% max. | |
0.806 to 0.810 (at 25°C) | |
C4H10O | |
MFCD00002964 | |
2.95 mPa/s at 20°C | |
0.1% max. | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12 | |
Certified ACS Reagent |
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Safety and Handling
WARNING!
Emergency Overview
Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid