missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Butoxy-2-propanol 96.0+%, TCI America™
$36.49 - $73.00
Chemical Identifiers
CAS | 5131-66-8 |
---|---|
Molecular Formula | C7H16O2 |
Molecular Weight (g/mol) | 132.203 |
MDL Number | MFCD00021904 |
InChI Key | RWNUSVWFHDHRCJ-UHFFFAOYSA-N |
Synonym | Propylene Glycol 1-Monobutyl Ether |
PubChem CID | 21210 |
IUPAC Name | 1-butoxypropan-2-ol |
SMILES | CCCCOCC(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B086425ML
|
TCI America
B086425ML |
25 mL |
Each for $36.49
|
|
|||||
B0864500ML
|
TCI America
B0864500ML |
500 mL |
Each for $73.00
|
|
|||||
Chemical Identifiers
5131-66-8 | |
132.203 | |
RWNUSVWFHDHRCJ-UHFFFAOYSA-N | |
21210 | |
CCCCOCC(C)O |
C7H16O2 | |
MFCD00021904 | |
Propylene Glycol 1-Monobutyl Ether | |
1-butoxypropan-2-ol |
Specifications
5131-66-8 | |
170°C | |
MFCD00021904 | |
Propylene Glycol 1-Monobutyl Ether | |
CCCCOCC(C)O | |
132.203 | |
132.20 | |
Liquid |
Colorless | |
C7H16O2 | |
25 mL | |
RWNUSVWFHDHRCJ-UHFFFAOYSA-N | |
1-butoxypropan-2-ol | |
21210 | |
≥96.0% (GC) | |
1-Butoxy-2-propanol |
Safety and Handling
EINECSNumber : (2)-2424&(7)-0097
RTECSNumber : UA7700000
TSCA : Yes