Learn More
1-Butylboronic acid, 98%, Thermo Scientific Chemicals
$61.73 - $668.38
Chemical Identifiers
CAS | 4426-47-5 |
---|---|
Molecular Formula | C4H11BO2 |
Molecular Weight (g/mol) | 101.94 |
MDL Number | MFCD00002106 |
InChI Key | QPKFVRWIISEVCW-UHFFFAOYSA-N |
Synonym | n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane |
PubChem CID | 20479 |
IUPAC Name | butylboronic acid |
SMILES | CCCCB(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1372503
|
Thermo Scientific Chemicals
A1372503 |
1 g |
Each for $61.73
|
|
|||||
AAA1372506
|
Thermo Scientific Chemicals
A1372506 |
5 g |
Each for $186.27
|
|
|||||
AAA1372514
|
Thermo Scientific Chemicals
A1372514 |
25 g |
Each for $668.38
|
|
|||||
Description
1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents. Store under dry inert gas.
Chemical Identifiers
4426-47-5 | |
101.94 | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
20479 | |
CCCCB(O)O |
C4H11BO2 | |
MFCD00002106 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
butylboronic acid |
Specifications
4426-47-5 | |
C4H11BO2 | |
1 g | |
Air sensitive; Hygroscopic | |
Soluble in water. | |
CCCCB(O)O | |
101.94 | |
101.94 | |
1-Butylboronic acid |
92°C to 96°C | |
MFCD00002106 | |
1733489 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
butylboronic acid | |
20479 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-607-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only