Learn More
1-Butylphosphonic acid, 98%, Thermo Scientific Chemicals
$93.30 - $1271.01
Chemical Identifiers
CAS | 3321-64-0 |
---|---|
Molecular Formula | C4H11O3P |
Molecular Weight (g/mol) | 138.10 |
MDL Number | MFCD00012295 |
InChI Key | UOKRBSXOBUKDGE-UHFFFAOYSA-N |
Synonym | n-butylphosphonic acid, 1-butanephosphonic acid, butylphosphonate, 1-butylphosphonic acid, phosphonic acid, butyl, n-butyl phosphonic acid, acmc-1cr41 |
PubChem CID | 76839 |
ChEBI | CHEBI:41384 |
IUPAC Name | butylphosphonic acid |
SMILES | CCCCP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1798303
|
Thermo Scientific Chemicals
A1798303 |
1 g |
Each for $93.30
|
|
|||||
AAA1798306
|
Thermo Scientific Chemicals
A1798306 |
5 g |
Each for $268.45
|
|
|||||
AAA1798314
|
Thermo Scientific Chemicals
A1798314 |
25 g |
Each for $1,271.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3321-64-0 | |
138.10 | |
UOKRBSXOBUKDGE-UHFFFAOYSA-N | |
76839 | |
butylphosphonic acid |
C4H11O3P | |
MFCD00012295 | |
n-butylphosphonic acid, 1-butanephosphonic acid, butylphosphonate, 1-butylphosphonic acid, phosphonic acid, butyl, n-butyl phosphonic acid, acmc-1cr41 | |
CHEBI:41384 | |
CCCCP(O)(O)=O |
Specifications
3321-64-0 | |
Odorless | |
MFCD00012295 | |
UN3261 | |
n-butylphosphonic acid, 1-butanephosphonic acid, butylphosphonate, 1-butylphosphonic acid, phosphonic acid, butyl, n-butyl phosphonic acid, acmc-1cr41 | |
CCCCP(O)(O)=O | |
138.10 | |
CHEBI:41384 | |
98% |
105°C to 107°C | |
C4H11O3P | |
1 g | |
1749026 | |
UOKRBSXOBUKDGE-UHFFFAOYSA-N | |
butylphosphonic acid | |
76839 | |
138.1 | |
1-Butylphosphonic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only