missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzene, 99%

CAS: 615-41-8 | C6H4ClI | 238.45 g/mol

$67.08 - $430.74

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1825509
View Documents
Thermo Scientific Chemicals
A1825509
10 g N/A
Only null left
 
AAA1825514
View Documents
Thermo Scientific Chemicals
A1825514
25 g N/A
Only null left
 
AAA1825522
View Documents
Thermo Scientific Chemicals
A1825522
100 g N/A
Only null left
 
Description

Description

1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2'-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2′-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place. Keep the container tightly closed.
Specifications

Chemical Identifiers

615-41-8
238.45
MPEOPBCQHNWNFB-UHFFFAOYSA-N
11993
ClC1=CC=CC=C1I
C6H4ClI
MFCD00001033
2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
1-chloro-2-iodobenzene

Specifications

615-41-8
1.954
111°C (231°F)
1.6345
10 g
Light sensitive
Miscible with most common organic solvents. Immiscible with water.
ClC1=CC=CC=C1I
238.45
238.46
1-Chloro-2-iodobenzene
1°C
108°C to 110°C (15 mmHg)
C6H4ClI
MFCD00001033
1905116
2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
MPEOPBCQHNWNFB-UHFFFAOYSA-N
1-chloro-2-iodobenzene
11993
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-423-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only