missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-iodoethane, 96%, stab. with copper, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1074214

 View more versions of this product

Catalog No. AAL1074214


Explore available promotions

Description

Description

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Chloro-2-iodoethane
2.088
>100°C (212°F)
1.566
25 g
Light sensitive
Soluble inorganic solvents. Insoluble in water.
ClCCI
190.41
190.41
624-70-4
61°C to 62°C (30 mmHg)
C2H4ClI
MFCD00041453
1730865
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
JTWWWQGSFTWWDL-UHFFFAOYSA-N
1-chloro-2-iodoethane
136438
96%

Chemical Identifiers

624-70-4
190.41
JTWWWQGSFTWWDL-UHFFFAOYSA-N
136438
ClCCI
C2H4ClI
MFCD00041453
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
1-chloro-2-iodoethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 677-827-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only