missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-methyl-2-propanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1897814

 View more versions of this product

Catalog No. AAA1897814


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chloro-2-methyl-2-propanol
1.058
38°C (100°F)
1.44
25 g
1697054
JNOZGFXJZQXOSU-UHFFFAOYSA-N
1-chloro-2-methylpropan-2-ol
68409
98%
558-42-9
124°C to 128°C
C4H9ClO
MFCD00021807
UN1987
1-chloro-2-methyl-2-propanol, chloro-tert-butanol, 2-propanol, 1-chloro-2-methyl, 1-chloro-tert-butyl alcohol, 2-chloro-tert-butyl alcohol, 1-chloro-2-methyl-propan-2-ol, pubchem3675, 1-chloro-2-isobutanol, acmc-209lpr, chloro-tert-butyl alcohol
CC(C)(O)CCl
108.57
108.57

Chemical Identifiers

558-42-9
108.57
JNOZGFXJZQXOSU-UHFFFAOYSA-N
68409
CC(C)(O)CCl
C4H9ClO
MFCD00021807
1-chloro-2-methyl-2-propanol, chloro-tert-butanol, 2-propanol, 1-chloro-2-methyl, 1-chloro-tert-butyl alcohol, 2-chloro-tert-butyl alcohol, 1-chloro-2-methyl-propan-2-ol, pubchem3675, 1-chloro-2-isobutanol, acmc-209lpr, chloro-tert-butyl alcohol
1-chloro-2-methylpropan-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 209-196-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only