missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals

$105.55 - $321.41

Chemical Identifiers

CAS 513-36-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000954
InChI Key QTBFPMKWQKYFLR-UHFFFAOYSA-N
PubChem CID 10554
IUPAC Name 1-chloro-2-methylpropane
SMILES CC(C)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0418118
View Documents
Thermo Scientific Chemicals
L0418118
50 g
Each for $105.55
 
AAL0418130
View Documents
Thermo Scientific Chemicals
L0418130
250 g
Each for $321.41
 
Description

Description

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

513-36-0
92.566
QTBFPMKWQKYFLR-UHFFFAOYSA-N
1-chloro-2-methylpropane
C4H9Cl
MFCD00000954
10554
CC(C)CCl

Specifications

513-36-0
0.879
−7°C (19°F)
C4H9Cl
MFCD00000954
UN1127
14,5138
Miscible with water,alcohol and diethyl ether.
CC(C)CCl
92.566
92.57
1-Chloro-2-methylpropane
-131°C
68°C to 69°C
Sweet
1.3975
50 g
635650
isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
QTBFPMKWQKYFLR-UHFFFAOYSA-N
1-chloro-2-methylpropane
10554
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

EINECSNumber : 208-157-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only