Learn More
1-Chloro-3,3-dimethylbutane, 97%
CAS: 2855-08-5 | C6H13Cl | 120.62 g/mol
Supplier: Thermo Scientific Chemicals L1175309
Description
1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Not miscible or difficult to mix in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications
| 1-Chloro-3,3-dimethylbutane | |
| 0.867 | |
| 7°C (44°F) | |
| 1.417 | |
| MFCD00018980 | |
| UN1993 | |
| neohexyl chloride, 3,3-dimethylbutyl chloride, butane, 1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethyl-butane, acmc-1clex, tert-butylchloroethylene, ksc561m9t, butane,1-chloro-3,3-dimethyl, 1-chloro-3,3-dimethylbutane 10g | |
| XGCKOSFYXBAPQM-UHFFFAOYSA-N | |
| 1-chloro-3,3-dimethylbutane | |
| 76111 | |
| 97% |
| 2855-08-5 | |
| 116°C to 118°C | |
| C6H13Cl | |
| (CH3)3CCH2CH2Cl | |
| 10 g | |
| 1731465 | |
| Not miscible or difficult to mix in water. | |
| CC(C)(C)CCCl | |
| 120.62 | |
| 120.62 |
Chemical Identifiers
| 2855-08-5 | |
| MFCD00018980 |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 220-665-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only