missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3,5-difluorobenzene 97.0+%, TCI America™
$74.56 - $275.87
Chemical Identifiers
| CAS | 1435-43-4 |
|---|---|
| Molecular Formula | C6H3ClF2 |
| Molecular Weight (g/mol) | 148.537 |
| MDL Number | MFCD00041518 |
| InChI Key | RFKBODCWHNDUTJ-UHFFFAOYSA-N |
| PubChem CID | 137000 |
| IUPAC Name | 1-chloro-3,5-difluorobenzene |
| SMILES | C1=C(C=C(C=C1F)Cl)F |
Chemical Identifiers
| 1435-43-4 | |
| 148.537 | |
| RFKBODCWHNDUTJ-UHFFFAOYSA-N | |
| 1-chloro-3,5-difluorobenzene |
| C6H3ClF2 | |
| MFCD00041518 | |
| 137000 | |
| C1=C(C=C(C=C1F)Cl)F |
Specifications
| 1435-43-4 | |
| 116°C | |
| MFCD00041518 | |
| 1993 | |
| RFKBODCWHNDUTJ-UHFFFAOYSA-N | |
| 1-chloro-3,5-difluorobenzene | |
| 137000 | |
| ≥97.0% (GC) | |
| 1-Chloro-3,5-difluorobenzene |
| Yellow | |
| C6H3ClF2 | |
| 5 g | |
| 3,5-difluorochlorobenzene, benzene, 1-chloro-3,5-difluoro, benzene,1-chloro-3,5-difluoro, 1-chloro-3,5-difluoro-benzene, 3,5-difluorochlorobenzene, 5-difluorochlorobenzene, pubchem4318, 3,5-difluorchlorbenzol, acmc-209cqr, intermediates-zcf02092, 1-chloro-3,5difluorobenzene | |
| C1=C(C=C(C=C1F)Cl)F | |
| 148.537 | |
| 148.54 | |
| Liquid |
Safety and Handling
TSCA : No