missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chloro-4-iodobutane (stabilized with Copper chip) 98.0+%, TCI America™
Supplier: TCI America C271325G
Specifications
1-Chloro-4-iodobutane (stabilized with Copper chip) | |
Yellow | |
C4H8ClI | |
25 g | |
JXOSPTBRSOYXGC-UHFFFAOYSA-N | |
1-chloro-4-iodobutane | |
82527 | |
≥98.0% (GC) |
10297-05-9 | |
67°C | |
MFCD00039415 | |
butane, 1-chloro-4-iodo, tetramethylene chloroiodide, 4-chlorobutyl iodide, 4-iodobutyl chloride, 1-chloro4-iodobutane, 4-chloro-1-iodobutane, 4-iodo-1-chlorobutane, 1-iodo-4-chlorobutane, acmc-1buxg, 1-chloro-4-iodo-butane | |
ClCCCCI | |
218.46 | |
218.46 | |
Liquid |
Chemical Identifiers
10297-05-9 | |
218.46 | |
JXOSPTBRSOYXGC-UHFFFAOYSA-N | |
82527 | |
ClCCCCI |
C4H8ClI | |
MFCD00039415 | |
butane, 1-chloro-4-iodo, tetramethylene chloroiodide, 4-chlorobutyl iodide, 4-iodobutyl chloride, 1-chloro4-iodobutane, 4-chloro-1-iodobutane, 4-iodo-1-chlorobutane, 1-iodo-4-chlorobutane, acmc-1buxg, 1-chloro-4-iodo-butane | |
1-chloro-4-iodobutane |
Safety and Handling
TSCA : Yes