Learn More
1-Chlorobutane, 99+%, Thermo Scientific Chemicals
$53.55 - $243.89
Chemical Identifiers
CAS | 109-69-3 |
---|---|
Molecular Formula | C4H9Cl |
Molecular Weight (g/mol) | 92.57 |
MDL Number | MFCD00001009 |
InChI Key | VFWCMGCRMGJXDK-UHFFFAOYSA-N |
Synonym | butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane |
PubChem CID | 8005 |
IUPAC Name | 1-chlorobutane |
SMILES | CCCCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA11494AE
|
Thermo Scientific Chemicals
A11494AE |
100 mL |
Each for $53.55
|
|
||||||
AAA114940F
|
Thermo Scientific Chemicals
A114940F |
2500 mL |
Each for $243.89
|
|
||||||
Description
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
109-69-3 | |
92.57 | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
8005 | |
CCCCCl |
C4H9Cl | |
MFCD00001009 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
1-chlorobutane |
Specifications
109-69-3 | |
0.886 | |
−12°C (10°F) | |
Pungent | |
1.4024 | |
100 mL | |
1730909 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
1-chlorobutane | |
8005 | |
≥99% | |
1-Chlorobutane |
-123°C | |
77°C to 78°C | |
>99% | |
C4H9Cl | |
MFCD00001009 | |
UN1127 | |
14,1560 | |
Miscible with alcohol and ether. Immiscible with water. | |
CCCCCl | |
92.57 | |
92.57 | |
Liquid |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber : 203-696-6
RTECSNumber : EJ6300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only