Learn More
1-Chlorobutane, 99+%, pure
CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
$176.15 - $1081.37
Identifiants chimiques
| CAS | 109-69-3 |
|---|---|
| Molecular Formula | C4H9Cl |
| Molecular Weight (g/mol) | 92.57 |
| MDL Number | MFCD00001009 |
| InChI Key | VFWCMGCRMGJXDK-UHFFFAOYSA-N |
| Synonym | butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane |
| PubChem CID | 8005 |
| IUPAC Name | 1-chlorobutane |
| SMILES | CCCCCl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC154630010
|
Thermo Scientific Chemicals
154630010 |
1 L | Glass bottle |
chaque for $176.15
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC154630025
|
Thermo Scientific Chemicals
154630025 |
2.5 L | Glass bottle |
chaque for $300.84
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC154630100
|
Thermo Scientific Chemicals
154630100 |
10 L | Plastic drum |
chaque for $1,081.37
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 109-69-3 | |
| 92.57 | |
| VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
| 8005 | |
| CCCCCl |
| C4H9Cl | |
| MFCD00001009 | |
| butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
| 1-chlorobutane |
Spécifications
| 109-69-3 | |
| 100.0 | |
| Colorless | |
| 77.0°C to 78.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4014 to 1.4034 | |
| MFCD00001009 | |
| 01, 118 | |
| 15, 1561 | |
| Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether | |
| CCCCCl | |
| 92.57 | |
| 0.45 mPa.s (20°C) | |
| 99+% | |
| Liquid |
| 99.0 | |
| -123.0°C | |
| 0.8800g/mL | |
| −12°C | |
| 99% min. (GC) | |
| C4H9Cl | |
| CH3(CH2)3Cl | |
| 1 L | |
| 0.88 | |
| butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
| VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
| 1-chlorobutane | |
| 8005 | |
| 92.57 | |
| Pure | |
| 1-Chlorobutane |
Sécurité et manipulation
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-696-6
RUO – Research Use Only