missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Chloromethyl)-1H-benzotriazole 98.0+%, TCI America™
$180.81 - $531.25
Chemical Identifiers
| CAS | 54187-96-1 |
|---|---|
| Molecular Formula | C7H6ClN3 |
| Molecular Weight (g/mol) | 167.60 |
| MDL Number | MFCD00216649 |
| InChI Key | VSEROABGEVRIRY-UHFFFAOYSA-N |
| PubChem CID | 3328222 |
| IUPAC Name | 1-(chloromethyl)-1H-1,2,3-benzotriazole |
| SMILES | ClCN1N=NC2=CC=CC=C12 |
Chemical Identifiers
| 54187-96-1 | |
| 167.60 | |
| VSEROABGEVRIRY-UHFFFAOYSA-N | |
| 1-(chloromethyl)-1H-1,2,3-benzotriazole |
| C7H6ClN3 | |
| MFCD00216649 | |
| 3328222 | |
| ClCN1N=NC2=CC=CC=C12 |
Specifications
| 54187-96-1 | |
| White-Yellow | |
| MFCD00216649 | |
| 3261 | |
| ClCN1N=NC2=CC=CC=C12 | |
| 167.60 | |
| 167.60 | |
| Crystalline Powder |
| 138°C | |
| C7H6ClN3 | |
| 5 g | |
| VSEROABGEVRIRY-UHFFFAOYSA-N | |
| 1-(chloromethyl)-1H-1,2,3-benzotriazole | |
| 3328222 | |
| ≥98.0% (T) | |
| 1-(Chloromethyl)-1H-benzotriazole |
Safety and Handling
TSCA : No