Learn More
1-Chlorophthalazin-4-one, 98%, Thermo Scientific Chemicals
$171.12 - $500.37
Chemical Identifiers
CAS | 2257-69-4 |
---|---|
Molecular Formula | C8H5ClN2O |
Molecular Weight (g/mol) | 180.59 |
MDL Number | MFCD00102192 |
InChI Key | QCKGMJDOJRNSMS-UHFFFAOYSA-N |
Synonym | 4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one |
PubChem CID | 2775142 |
SMILES | ClC1=NNC(=O)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2751503
|
Thermo Scientific Chemicals
H2751503 |
1 g |
Each for $171.12
|
|
|||||
AAH2751506
|
Thermo Scientific Chemicals
H2751506 |
5 g |
Each for $500.37
|
|
|||||
Description
1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.
Notes
Incompatible with oxidizing agents and bases.
Chemical Identifiers
2257-69-4 | |
180.59 | |
QCKGMJDOJRNSMS-UHFFFAOYSA-N | |
2775142 |
C8H5ClN2O | |
MFCD00102192 | |
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one | |
ClC1=NNC(=O)C2=CC=CC=C12 |
Specifications
2257-69-4 | |
C8H5ClN2O | |
1 g | |
QCKGMJDOJRNSMS-UHFFFAOYSA-N | |
4-chloro-1,2-dihydrophthalazin-1-one | |
2775142 | |
98% |
270°C to 274°C | |
MFCD00102192 | |
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one | |
ClC1=NNC(=O)C2=CC=CC=C12 | |
180.59 | |
180.59 | |
1-Chlorophthalazin-4-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only