missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chlorotriacontane 85.0+%, TCI America™
Supplier: TCI America T1911100MG
Specifications
1-Chlorotriacontane | |
66°C | |
C30H61Cl | |
100 mg | |
NSPRLRIWTSAZOP-UHFFFAOYSA-N | |
1-chlorotriacontane | |
15611905 | |
≥85.0% (GC) |
62016-82-4 | |
White | |
MFCD03093643 | |
Triacontyl Chloride | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCl | |
457.268 | |
457.27 | |
Crystalline Powder |
Chemical Identifiers
62016-82-4 | |
457.268 | |
NSPRLRIWTSAZOP-UHFFFAOYSA-N | |
15611905 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCl |
C30H61Cl | |
MFCD03093643 | |
Triacontyl Chloride | |
1-chlorotriacontane |
Safety and Handling
TSCA : No