missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclopentylethanol, 97%
CAS: 52829-98-8 | C7H14O | 114.19 g/mol
$148.64 - $540.18
Chemical Identifiers
| CAS | 52829-98-8 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.19 |
| MDL Number | MFCD00013743 |
| InChI Key | WPCMSUSLCWXTKB-UHFFFAOYNA-N |
| Synonym | 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl |
| PubChem CID | 95434 |
| IUPAC Name | 1-cyclopentylethanol |
| SMILES | CC(O)C1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAL0344604
|
Thermo Scientific Chemicals
L0344604 |
2 g |
Each for $148.64
|
|
|||||
|
AAL0344609
|
Thermo Scientific Chemicals
L0344609 |
10 g |
Each for $540.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 52829-98-8 | |
| 114.19 | |
| WPCMSUSLCWXTKB-UHFFFAOYNA-N | |
| 95434 | |
| CC(O)C1CCCC1 |
| C7H14O | |
| MFCD00013743 | |
| 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl | |
| 1-cyclopentylethanol |
Specifications
| 52829-98-8 | |
| 68°C (8 mmHg) | |
| 1.458 | |
| 2 g | |
| 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl | |
| CC(O)C1CCCC1 | |
| 114.19 | |
| 114.19 | |
| 1-Cyclopentylethanol |
| 0.911 | |
| C7H14O | |
| MFCD00013743 | |
| 2069851 | |
| WPCMSUSLCWXTKB-UHFFFAOYNA-N | |
| 1-cyclopentylethanol | |
| 95434 | |
| 97% |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 258-208-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only