Learn More
1-Decanethiol, 96%, Thermo Scientific Chemicals
$67.58 - $708.56
Chemical Identifiers
CAS | 143-10-2 |
---|---|
Molecular Formula | C10H22S |
Molecular Weight (g/mol) | 174.346 |
MDL Number | MFCD00004884 |
InChI Key | VTXVGVNLYGSIAR-UHFFFAOYSA-N |
Synonym | 1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol |
PubChem CID | 8917 |
IUPAC Name | decane-1-thiol |
SMILES | CCCCCCCCCCS |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA15913AC
|
Thermo Scientific Chemicals
A15913AC |
25 mL |
Each for $67.58
|
|
|||||
AAA15913AE
|
Thermo Scientific Chemicals
A15913AE |
100 mL |
Each for $182.07
|
|
|||||
AAA15913AP
|
Thermo Scientific Chemicals
A15913AP |
500 mL |
Each for $708.56
|
|
|||||
Description
1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. It is sensitive to air. Incompatible with strong oxidizing agents.
Chemical Identifiers
143-10-2 | |
174.346 | |
VTXVGVNLYGSIAR-UHFFFAOYSA-N | |
8917 | |
CCCCCCCCCCS |
C10H22S | |
MFCD00004884 | |
1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol | |
decane-1-thiol |
Specifications
143-10-2 | |
0.843 | |
98°C (208°F) | |
C10H22S | |
CH3(CH2)9SH | |
25 mL | |
Air Sensitive | |
Insoluble in water. | |
CCCCCCCCCCS | |
174.346 | |
174.35 | |
1-Decanethiol |
-26°C | |
237°C | |
Unpleasant | |
1.4565 | |
MFCD00004884 | |
1735223 | |
1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol | |
VTXVGVNLYGSIAR-UHFFFAOYSA-N | |
decane-1-thiol | |
8917 | |
96% |
Safety and Handling
GHS H Statement
H318-H335
Causes serious eye damage.
May cause respiratory irritation.
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
EINECSNumber : 205-584-2
TSCA : Yes
Recommended Storage : Store under Argon
RUO – Research Use Only