Learn More
1-Decanol, 98+%
CAS: 112-30-1 | C10H22O | 158.285 g/mol
Supplier: Thermo Scientific Chemicals A17288AP
Description
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications
| 1-Decanol | |
| 6°C | |
| 230°C to 232°C | |
| >98% | |
| 1.437 | |
| MFCD00004747 | |
| UN3082 | |
| 14,2855 | |
| Soluble in alcohol,ether; Insoluble in water;Miscible with carbon tetrachloride,ethanol,ether,acetone,benzene,chloroform and glacial acetic acid; Immiscible with water. | |
| CCCCCCCCCCO | |
| 158.285 | |
| CHEBI:28903 | |
| ≥98% |
| 112-30-1 | |
| 0.828 g/mL | |
| 82°C (179°F) | |
| C10H22O | |
| CH3(CH2)8CH2OH | |
| 500 mL | |
| 1735221 | |
| 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac | |
| MWKFXSUHUHTGQN-UHFFFAOYSA-N | |
| decan-1-ol | |
| 8174 | |
| 158.29 |
Chemical Identifiers
| 112-30-1 | |
| 158.285 | |
| MWKFXSUHUHTGQN-UHFFFAOYSA-N | |
| 8174 | |
| decan-1-ol |
| C10H22O | |
| MFCD00004747 | |
| 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac | |
| CHEBI:28903 | |
| CCCCCCCCCCO |
Safety and Handling
GHS H Statement
H332-H315-H319-H227
P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
H227-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 203-956-9
RTECSNumber : HE4375000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only