Learn More
1-Dimethylamino-2-propyne, 98%, Thermo Scientific Chemicals
$58.64 - $193.67
Chemical Identifiers
CAS | 7223-38-3 |
---|---|
Molecular Formula | C5H9N |
Molecular Weight (g/mol) | 83.134 |
MDL Number | MFCD00008575 |
InChI Key | ILBIXZPOMJFOJP-UHFFFAOYSA-N |
Synonym | 3-dimethylamino-1-propyne, 1-dimethylamino-2-propyne, 2-propyn-1-amine, n,n-dimethyl, n,n-dimethylpropargylamine, dimethyl prop-2-yn-1-yl amine, n,n-dimethyl-2-propyn-1-amine, dimethyl prop-2-ynyl amine, n,n-dimethyl propargylamine, n,n-dimethyl-2-propynylamine, dimethylpropargylamine |
PubChem CID | 81643 |
IUPAC Name | N,N-dimethylprop-2-yn-1-amine |
SMILES | CN(C)CC#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1233606
|
Thermo Scientific Chemicals
L1233606 |
5 g |
Each for $58.64
|
|
|||||
AAL1233614
|
Thermo Scientific Chemicals
L1233614 |
25 g |
Each for $193.67
|
|
|||||
Description
1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to carbon dioxide. Store under inert gas.
Chemical Identifiers
7223-38-3 | |
83.134 | |
ILBIXZPOMJFOJP-UHFFFAOYSA-N | |
81643 | |
CN(C)CC#C |
C5H9N | |
MFCD00008575 | |
3-dimethylamino-1-propyne, 1-dimethylamino-2-propyne, 2-propyn-1-amine, n,n-dimethyl, n,n-dimethylpropargylamine, dimethyl prop-2-yn-1-yl amine, n,n-dimethyl-2-propyn-1-amine, dimethyl prop-2-ynyl amine, n,n-dimethyl propargylamine, n,n-dimethyl-2-propynylamine, dimethylpropargylamine | |
N,N-dimethylprop-2-yn-1-amine |
Specifications
7223-38-3 | |
80°C to 81°C | |
C5H9N | |
MFCD00008575 | |
UN2733 | |
Air Sensitive | |
Fully miscible in water. | |
CN(C)CC#C | |
83.134 | |
83.13 | |
1-Dimethylamino-2-propyne |
0.772 | |
−7°C (19°F) | |
1.42 | |
5 g | |
1071222 | |
3-dimethylamino-1-propyne, 1-dimethylamino-2-propyne, 2-propyn-1-amine, n,n-dimethyl, n,n-dimethylpropargylamine, dimethyl prop-2-yn-1-yl amine, n,n-dimethyl-2-propyn-1-amine, dimethyl prop-2-ynyl amine, n,n-dimethyl propargylamine, n,n-dimethyl-2-propynylamine, dimethylpropargylamine | |
ILBIXZPOMJFOJP-UHFFFAOYSA-N | |
N,N-dimethylprop-2-yn-1-amine | |
81643 | |
98% |
Safety and Handling
GHS H Statement
H225-H301-H311-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 230-620-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only