missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Docosanol, 98%
CAS: 661-19-8 | C22H46O | 326.61 g/mol
$86.61 - $86.61
Chemical Identifiers
| CAS | 661-19-8 |
|---|---|
| Molecular Formula | C22H46O |
| Molecular Weight (g/mol) | 326.61 |
| MDL Number | MFCD00002939 |
| InChI Key | NOPFSRXAKWQILS-UHFFFAOYSA-N |
| Synonym | 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 |
| PubChem CID | 12620 |
| ChEBI | CHEBI:31000 |
| IUPAC Name | docosan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC173720250
|
Thermo Scientific Chemicals
173720250 |
25 g | Plastic bottle |
Each for $86.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 661-19-8 | |
| 326.61 | |
| NOPFSRXAKWQILS-UHFFFAOYSA-N | |
| 12620 | |
| docosan-1-ol |
| C22H46O | |
| MFCD00002939 | |
| 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 | |
| CHEBI:31000 | |
| CCCCCCCCCCCCCCCCCCCCCCO |
Specifications
| 661-19-8 | |
| 100 | |
| White | |
| Authentic | |
| Plastic bottle | |
| CH3(CH2)21OH | |
| 25 g | |
| 15, 3444 | |
| Solubility in water: insoluble. | |
| CCCCCCCCCCCCCCCCCCCCCCO | |
| 326.61 | |
| 7.5 mPa.s (80°C) | |
| 326.61 | |
| Pellets or Tablets |
| 97.5 | |
| 68.0°C to 72.0°C | |
| 227°C | |
| 97.5% min. (GC) | |
| C22H46O | |
| MFCD00002939 | |
| 01, 431 | |
| 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22 | |
| NOPFSRXAKWQILS-UHFFFAOYSA-N | |
| docosan-1-ol | |
| 12620 | |
| CHEBI:31000 | |
| 98% | |
| 1-Docosanol, 0.98 |
Safety and Handling
EINECSNumber : 211-546-6
RUO – Research Use Only