Learn More
1-Dodecanesulfonyl chloride, 97%, Thermo Scientific Chemicals
$155.59 - $504.03
Chemical Identifiers
CAS | 10147-40-7 |
---|---|
Molecular Formula | C12H25ClO2S |
Molecular Weight (g/mol) | 268.84 |
MDL Number | MFCD00014728 |
InChI Key | PBULHKIPTBIZHO-UHFFFAOYSA-N |
Synonym | 1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chloride |
PubChem CID | 66280 |
IUPAC Name | dodecane-1-sulfonyl chloride |
SMILES | CCCCCCCCCCCCS(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0122406
|
Thermo Scientific Chemicals
L0122406 |
5 g |
Each for $155.59
|
|
||||||
AAL0122414
|
Thermo Scientific Chemicals
L0122414 |
25 g |
Each for $504.03
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10147-40-7 | |
268.84 | |
PBULHKIPTBIZHO-UHFFFAOYSA-N | |
66280 | |
CCCCCCCCCCCCS(Cl)(=O)=O |
C12H25ClO2S | |
MFCD00014728 | |
1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chloride | |
dodecane-1-sulfonyl chloride |
Specifications
10147-40-7 | |
126°C to 130°C (4 mmHg) | |
CH3(CH2)11SO2Cl | |
5 g | |
1778092 | |
1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chloride | |
CCCCCCCCCCCCS(Cl)(=O)=O | |
268.84 | |
268.84 | |
1-Dodecanesulfonyl chloride |
39°C to 42°C | |
C12H25ClO2S | |
MFCD00014728 | |
UN3261 | |
Moisture sensitive | |
PBULHKIPTBIZHO-UHFFFAOYSA-N | |
dodecane-1-sulfonyl chloride | |
66280 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 233-416-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only