Learn More
1-Dodecylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A15515A1
Description
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
Solubility
Soluble in water (78 mg/L 25°C).
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications
1-Dodecylamine | |
27°C to 31°C | |
64°C to 66°C (0.05 mmHg) | |
≥98% | |
MFCD00008154 | |
UN3259 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
dodecan-1-amine | |
13583 | |
≥98% |
124-22-1 | |
0.802 | |
115°C (239°F) | |
C12H27N | |
1 kg | |
1633576 | |
Soluble in water (78mg/L 25°C). | |
CCCCCCCCCCCCN | |
185.36 | |
185.36 |
Chemical Identifiers
124-22-1 | |
185.36 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
13583 | |
CCCCCCCCCCCCN |
C12H27N | |
MFCD00008154 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
dodecan-1-amine |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H304-H314-H335-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 204-690-6
RTECSNumber : JR6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only