Learn More
1-Ethoxy-2-propanol, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 259050100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Ethoxy-2-propanol | |
0.8970g/mL | |
42°C | |
94% min. Remark: balance 2-ethoxy-1-propanol (GC) | |
C5H12O2 | |
C2H5OCH2CH(OH)CH3 | |
10L | |
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether | |
JOLQKTGDSGKSKJ-UHFFFAOYNA-N | |
1-ethoxypropan-2-ol | |
15287 | |
104.14 |
1569-02-4 | |
132°C | |
Authentic | |
Plastic drum | |
1.4050 to 1.409 | |
MFCD00067050 | |
0.897 | |
Solubility in water: soluble | |
CCOCC(C)O | |
104.15 | |
2 mPa.s (20°C) | |
95% |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 216-374-5
RUO â Research Use Only