missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 95%

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol

$190.43 - $341.84

Chemical Identifiers

CAS 1569-02-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00067050
InChI Key JOLQKTGDSGKSKJ-UHFFFAOYNA-N
Synonym 1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
PubChem CID 15287
IUPAC Name 1-ethoxypropan-2-ol
SMILES CCOCC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259050010
SDS View Documents
Thermo Scientific Chemicals
259050010
1 L Glass Bottle
Each for $190.43
Only null left
 
AC259050025
SDS View Documents
Thermo Scientific Chemicals
259050025
2.5 L Glass Bottle
Each for $341.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1569-02-4
104.15
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
15287
CCOCC(C)O
C5H12O2
MFCD00067050
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
1-ethoxypropan-2-ol

Specifications

1569-02-4
Colorless
132°C
Authentic
Glass Bottle
1.4050 to 1.409
MFCD00067050
0.897
Solubility in water: soluble
CCOCC(C)O
104.15
2 mPa.s (20°C)
95%
1-Ethoxy-2-propanol
-100°C
0.8970g/mL
42°C
94% min. Remark: balance 2-ethoxy-1-propanol (GC)
C5H12O2
C2H5OCH2CH(OH)CH3
1 L
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
1-ethoxypropan-2-ol
15287
104.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 216-374-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only