missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-1-methylpyrrolidinium bromide, 98%, Thermo Scientific Chemicals

$175.14 - $478.45

Chemical Identifiers

CAS 69227-51-6
Molecular Formula C7H16BrN
Molecular Weight (g/mol) 194.116
MDL Number MFCD03095384
InChI Key KHJQQUGSPDBDRM-UHFFFAOYSA-M
Synonym 1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
PubChem CID 112264
IUPAC Name 1-ethyl-1-methylpyrrolidin-1-ium;bromide
SMILES CC[N+]1(CCCC1)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6605014
View Documents
Thermo Scientific Chemicals
H6605014
25 g
Each for $175.14
 
AAH6605022
View Documents
Thermo Scientific Chemicals
H6605022
100 g
Each for $478.45
 
Description

Description

1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

69227-51-6
194.116
KHJQQUGSPDBDRM-UHFFFAOYSA-M
112264
CC[N+]1(CCCC1)C.[Br-]
C7H16BrN
MFCD03095384
1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
1-ethyl-1-methylpyrrolidin-1-ium;bromide

Specifications

69227-51-6
MFCD03095384
14507266
Soluble in water.
CC[N+]1(CCCC1)C.[Br-]
194.116
194.12
Solid
C7H16BrN
25 g
1-ethyl-1-methylpyrrolidinium bromide, 1-ethyl-1-methylpyrrolidin-1-ium bromide, 1-ethyl-1-methylpyrrolidiniumbromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide, 1-methyl-1-ethylpyrrolidinium bromide, pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1, dsstox_cid_29306, dsstox_rid_83422, dsstox_gsid_49347, ksc495s4d
KHJQQUGSPDBDRM-UHFFFAOYSA-M
1-ethyl-1-methylpyrrolidin-1-ium;bromide
112264
98%
1-Ethyl-1-methylpyrrolidinium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P281-P308+P313-P501c

H341

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only