missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals

$117.05 - $763.34

Chemical Identifiers

CAS 606-55-3
Molecular Formula C12H14IN
Molecular Weight (g/mol) 299.155
MDL Number MFCD00031829
InChI Key OEVSHJVOKFWBJY-UHFFFAOYSA-M
Synonym 1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
PubChem CID 69076
IUPAC Name 1-ethyl-2-methylquinolin-1-ium;iodide
SMILES CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1058406
View Documents
Thermo Scientific Chemicals
A1058406
5 g
Each for $117.05
 
AAA1058414
View Documents
Thermo Scientific Chemicals
A1058414
25 g
Each for $304.97
 
AAA1058422
View Documents
Thermo Scientific Chemicals
A1058422
100 g
Each for $763.34
 
Description

Description

1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

Solubility
Soluble in Water.

Notes
Precursor in synthesis of carbocyanine dyes.
Specifications

Chemical Identifiers

606-55-3
299.155
OEVSHJVOKFWBJY-UHFFFAOYSA-M
69076
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]
C12H14IN
MFCD00031829
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
1-ethyl-2-methylquinolin-1-ium;iodide

Specifications

606-55-3
C12H14IN
5 g
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
OEVSHJVOKFWBJY-UHFFFAOYSA-M
1-ethyl-2-methylquinolin-1-ium;iodide
69076
97%
231°C (decomposition)
MFCD00031829
Light sensitive
Soluble in Water.
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]
299.155
299.16
1-Ethyl-2-methylquinolinium iodide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-120-7

RTECSNumber : VA0740000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only